Learn More
Liebherr™ GN Series Freezer
Use these freezers for general laboratory storage of temperature-sensitive samples. Liebherr™ GN Series Freezers provide expert NoFrost refrigeration technology to ensure long-term freshness.
1155.00€
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
12527066
|
Liebherr™
GN 3113 |
307 L |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on 056 260 260 for assistance. |
||||||
Description
Automatic SuperFrost function
- Makes freezing simple and saves energy
- quickly reduces the temperature to -32 °C and thus creates the cold air required to preserve vital vitamins
- As soon as the food is completely frozen, the automatic SuperFrost function switches back to normal mode after max. 65 hours, and thus assists in saving electricity
FrostSafe
- With the FrostSafe, the extra-high and removable drawers are closed all round
- Cold cannot dissipate so quickly when the appliance is opened
- Transparent front of the compartments guarantees an optimum overview of the frozen items
VarioSpace
- All freezers with NoFrost and SmartFrost feature drawers and intermediate glass shelves underneath them which can be conveniently removed
- This results in VarioSpace; the practical system for extra storage space so that space can be created quickly even for larger frozen food items
Slimline door handle with integrated opening mechanism
- The practical door handles with integrated opening mechanism ensure that the appliance doors can be opened with a minimum of effort even if they are opened several times in rapid succession
- Handle mechanism assists in pushing the door from the appliance housing
Specifications
10°C to 43°C | |
White | |
R600A |