Learn More
GNA15 Monoclonal Antibody (OTI1D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA808159S
Description
GNA15 Monoclonal antibody specifically detects GNA15 in Human samples. It is validated for Western BlotSpecifications
| GNA15 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P36888 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2322 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1D3 | |
| Unconjugated | |
| FLT3 | |
| Epididymis tissue protein Li 17E; G alpha 15; G alpha 16; g alpha-15; G alpha-16; G protein subunit alpha 15; G[a]15; Galpha15; GNA 15; GNA 16; GNA15; Gna-15; GNA16; G-protein subunit alpha-15; G-protein subunit alpha-16; Gq class; guanine nucleotide binding protein (G protein), alpha 15 (Gq class); guanine nucleotide binding protein, alpha 15; guanine nucleotide-binding protein subunit alpha-15; Guanine nucleotide-binding protein subunit alpha-16 | |
| Human recombinant protein fragment corresponding to amino acids 169-444 of human FLT3 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.