missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNG4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13780-1-AP-20UL
This item is not returnable.
View return policy
Description
Guanine nucleotide-binding proteins (G proteins) are involved as a modulator or transducer in various transmembrane signaling systems. The beta and gamma chains are required for the GTPase activity, for replacement of GDP by GTP, and for G protein-effector interaction.Specifications
| GNG4 | |
| Polyclonal | |
| Unconjugated | |
| GNG4 | |
| G gamma4, GNG4, GNGT4 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 114118, 14706, 2786 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P50150, P50153 | |
| GNG4 | |
| GNG4 Fusion Protein Ag4787 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction