Learn More
GNS Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13044-1-AP-150UL
 View more versions of this product
Product Code. 16811064
  • 410.00€ / 150µL
Description
GNS is a lysosomal enzyme found in all cells. It is involved in the catabolism of heparin, heparan sulphate, and keratan sulphate. Deficiency of this enzyme results in the accumulation of undegraded substrate and the lysosomal storage disorder mucopolysaccharidosis type IIID (Sanfilippo D syndrome). Mucopolysaccharidosis type IIID is the least common of the four subtypes of Sanfilippo syndrome.
Specifications
Specifications
GNS
Polyclonal
Unconjugated
GNS
G6S, Glucosamine 6 sulfatase, GNS
Rabbit
Antigen Affinity Chromatography
RUO
2799, 299825, 75612
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.32 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P15586, Q8BFR4
GNS
GNS Fusion Protein Ag3712
150 μL