missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Goat anti-Canine IgG (H+L) Secondary Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP2-60652-0.5mg
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal specifically detects IgG (H+L) in Canine samples. It is validated for Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunomicroscopy.
Specifications
| IgG (H+L) | |
| Polyclonal | |
| Unconjugated | |
| 10 mM Sodium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.05% Sodium Azide | |
| Purified Dog IgG (H&L) | |
| 0.5 mg | |
| Secondary | |
| Store at 4°C. |
| Western Blot, Immunocytochemistry, Immunofluorescence, Immunomicroscopy | |
| 4.5mg/mL | |
| Flow Cytometry 1:20-1:2000, Immunocytochemistry/Immunofluorescence, Immunomicroscopy 1:20-1:2000 | |
| Goat | |
| Affinity purified | |
| Adaptive Immunity, Immunology | |
| Based on IEP, this antibody reacts with heavy gamma chains on dog IgG and light chains on all dog immunoglobulins | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction