missing translation for 'onlineSavingsMsg'
Learn More
Learn More
golgin 97 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12640-1-AP-20UL
This item is not returnable.
View return policy
Description
The MW of this protein is 97 kDa, and 12640-1-AP specifically recognizes the 97 kDa protein. IF result shows strong Golgi staining.
The Golgi apparatus, which participates in glycosylation and transport of proteins and lipids in the secretory pathway, consists of a series of stacked cisternae (flattened membrane sacs). Interactions between the Golgi and microtubules are thought to be important for the reorganization of the Golgi after it fragments during mitosis. The Golgins are a family of proteins, of which the protein encoded by this gene is a member, that are localized to the Golgi. This encoded protein is associated with Sjogren′s syndrome.Specifications
| golgin 97 | |
| Polyclonal | |
| Unconjugated | |
| GOLGA1 | |
| GOLGA1, Golgin 97, Golgin subfamily A member 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 2800, 76899 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunoprecipitation, Immunocytochemistry | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q92805, Q9CW79 | |
| Golga1 | |
| Golgin 97 Fusion Protein Ag3335 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction