Learn More
Thermo Scientific™ GoPure™ MabCaptureC™ Screening Plate, 96-well, 20 μL
Shop All Thermo Scientific ProductsDescription
GoPure MabCaptureC™ 96-well screening plates contain MabCaptureC Affinity Matrix and provide a versatile platform for sample preparation, handling, and analysis. They accommodate multiple samples simultaneously to achieve high throughput and data generation.
These pre-filled screening plates have the following features:
- High-throughput screening capability enabling multiple conditions to be evaluated in a single plate experiment
- 20 μL per well, helping preserve sample while maximizing data generation
- Ready-to-use 96-well format for efficient evaluation of Thermo Scientific resins
Specifications
Specifications
| Concentration | 220 μL Resin Slurry/Well |
| Content And Storage | Store at 2°C to 8°C. Do not freeze. |
| Shipping Condition | Wet Ice |
| Particle Size | 70 μm |
| Form | Resin Slurry |
| Product Line | MabCaptureC |
| Quantity | 1 Plate |
| For Use With (Application) | Chromatography |
| Type | Affinity Screening Plate |
| Sample Volume | 20 μL |
| Show More |