missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GORAB Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17798-1-AP-20UL
This item is not returnable.
View return policy
Description
TCEB1 is known as protein elongin C, which is a subunit of the transcription factor B (SIII) complex. The SIII complex is composed of elongins A/A2, B and C. It activates elongation by RNA polymerase II by suppressing transient pausing of the polymerase at many sites within transcription units. Elongin A functions as the transcriptionally active component of the SIII complex, whereas elongins B and C are regulatory subunits. Elongin A2 is specifically expressed in the testis, and capable of forming a stable complex with elongins B and C. The von Hippel-Lindau tumor suppressor protein binds to elongins B and C, and thereby inhibits transcription elongation. Western blots using two different antibodies (P100962-T200 and P100963_P050 / P100963_P100) against two unique regions of this protein target confirm the same apparent molecular weight in our tests.Specifications
| GORAB | |
| Polyclonal | |
| Unconjugated | |
| GORAB | |
| GO, Golgin, RAB6 interacting, GORAB, hNTKL BP1, NTKL binding protein 1, NTKL BP1, NTKLBP1, RAB6 interacting Golgin, SCY1 like 1 binding protein 1, SCYL1 binding protein 1, SCYL1 BP1, SCYL1BP1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 304923, 92344, 98376 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| B1H222, Q5T7V8, Q8BRM2 | |
| GORAB | |
| GORAB Fusion Protein Ag12002 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction