Learn More
GORASP1 Monoclonal Antibody (OTI4F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501392
Description
Grasp65 is a structural protein of the golgi apparatus. It has been implicated in the stacking of golgi cisternae. Grasp65 is shown to have the properties to bind surfaces together in a mitotically regulated matter.Specifications
| GORASP1 | |
| Monoclonal | |
| 0.98 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQQ3 | |
| GORASP1 | |
| Full length human recombit protein of human GORASP1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
| Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI4F8 | |
| Unconjugated | |
| GORASP1 | |
| GOLPH5, GRASP65, P65 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 64689 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.