missing translation for 'onlineSavingsMsg'
Learn More

GOSR1 Antibody (HFD9), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97437-0.2MG

 View more versions of this product

Product Code. 18017729

  • 799.00€ / 0.20mg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

GOSR1 Monoclonal specifically detects GOSR1 in Mouse, Rat, Porcine, Bovine, Canine, Chicken, Rabbit, Sheep samples. It is validated for Western Blot, Electron Microscopy.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

GOSR1
Monoclonal
Unconjugated
PBS (pH 7.2) and 50% Glycerol with 0.09% Sodium Azide
GOSR1
Rat liver Golgi-enriched membranes.
0.2 mg
Membrane Trafficking and Chaperones
9527
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Western Blot, Immunoassay
HFD9
Western Blot 1:1000/ECL, Electron Microscopy 1:10-1:500
golgi integral membrane protein 2, golgi SNAP receptor complex member 1,28 kDa cis-Golgi SNARE p28, Golgi SNARE 28 kDa, GOLIM2, GOS28cis-golgi SNARE, GOS-28GOS28/P28, GS2828 kDa Golgi SNARE protein, P28
Mouse
Protein G purified
RUO
Primary
Mouse, Rat, Pig, Bovine, Canine, Chicken, Rabbit, Sheep
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only