Learn More
Gosselin™ Natural Polystyrene Flat Base Tube
Product Code. 11783303
Change view
Click to view available options
Bottom Shape:
Flat
Unit Size:
Pack of 1000
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Bottom Shape Unit Size
11783303 Flat Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11783303 Supplier Gosselin Supplier No. FLY3503
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
Specifications
Bottom Shape Flat
Capacity (Metric) 35 mL
Color Natural
For Use With (Application) Clinical, dairy, environment, industries, microbiology
Height (Metric) 95 mm