Learn More
Corning™ Straw Pipets, Polypropylene, Sterile
GREENER_CHOICE
Product Code. 12602255
Change view
Click to view available options
Volume (Metric):
1 mL
2.2 mL
5 mL
Length (Metric):
7.3 cm
19 cm
24 cm
Packaging Type:
10/Bag
100/Bag
25/Bag
40 Bags/25 Units
50/Bag
Quantity:
100/Bag
1000
3600/Cs.
6000/Cs.
Unit Size:
Pack of 1000
Pack of 12000
Pack of 3600
Pack of 6000
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Length (Metric) Packaging Type Quantity unitSize
12602255 2.2 mL 24 cm 50/Bag 6000/Cs. Pack of 6000
11562772 1 mL 19 cm 40 Bags/25 Units 1000 Pack of 1000
11992815 1 mL 19 cm 40 Bags/25 Units 1000 Pack of 12000
11825380 2.2 mL 24 cm 25/Bag 6000/Cs. Pack of 6000
12672275 5 mL 7.3 cm 10/Bag 3600/Cs. Pack of 3600
11962785 1 mL 19 cm 100/Bag 100/Bag Pack of 12000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 12602255 Supplier Corning™ Supplier No. EPI24B02
Estimated Shipment: 17-12-2026
Add to basket
Add to basket
Decrease cross contamination risks using disposable pipets
  • Product traceability on each bag
  • Sterile 10-3
  • Packaged individually in tamper evident single zip bags
TRUSTED_SUSTAINABILITY
Specifications
Material Polypropylene
Sterility Sterile
Quantity 6000/Cs.
Packaging Type 50/Bag
Volume (Metric) 2.2 mL
Length (Metric) 24 cm
Type Straw Pipet
Disposable Single-use