Learn More
A&D Weighing GP Series Precision Balances Internal Calibration Class II Approved
GREENER_CHOICE
Product Code. 16601132
Change view
Click to view available options
Capacity:
101 kg
101 kg/61 kg
12 kg
21 kg
31 kg
31 kg/6.1 kg
61 kg
Readability:
0.1 g
1 g
1 g/0.1 g (smart range)
10 g/1 g (smart range)
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Readability unitSize
16601132 31 kg 0.1 g Each
16621132 31 kg/6.1 kg 1 g/0.1 g (smart range) Each
16651122 101 kg 1 g Each
16651132 61 kg 1 g Each
16671122 101 kg/61 kg 10 g/1 g (smart range) Each
16681122 12 kg 0.1 g Each
16691122 21 kg 0.1 g Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Regulatory: This product is under eco-contribution tax.
Product Code. 16601132 Supplier A&D Weighing Supplier No. GP30KEC
  • ONLINE EXCLUSIVE PRICE
    4887.75€
    4535.35€ / Pack of 1
    Save 352.40€
    7% Off
Estimated Shipment: 24-09-2026
Add to basket
Add to basket
Regulatory: This product is under eco-contribution tax.
With capacities from 12 to 102 kg, these high precision industrial balances have a large weighing pan, and also feature IP65 dust and waterproof protection. SmartRange feature for improved precision, regardless of tare container weight.
  • EC Type approved
  • Swing arm design or separate display and platform design
  • IP65 protected
  • Durable aluminum alloy & stainless steel construction
  • Easy calibration with internal calibration mass
  • SHS response speed of only 1.5 seconds
  • GLP, GMP and ISO compliant
  • RS-232C interface
  • Comparator output and analog output (optional)
  • Large Vacuum Fluorescent display
  • Data memory function
  • Counting function with ACAI (Automatic Counting Accuracy Improvement)
  • Smart range function (GP-32K/102K/32KS)
  • Comparator function
  • Underhook weighing for thermal and magnetic samples as well as for density calculation
  • Animal weighing function
Specifications
Capacity 31 kg
Display Type VFD
Readability 0.1 g
Stabilization Time 1.5 seconds
Type Industrial Precision Balance