missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP130 Monoclonal Antibody (125604)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA561471
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
The protein encoded by this gene is a signal transducer shared by many cytokines, including interleukin 6 (IL6), ciliary neurotrophic factor (CNTF), leukemia inhibitory factor (LIF), and oncostatin M (OSM). This protein functions as a part of the cytokine receptor complex. The activation of this protein is dependent upon the binding of cytokines to their receptors. vIL6, a protein related to IL6 and encoded by the Kaposi sarcoma-associated herpesvirus, can bypass the interleukin 6 receptor (IL6R) and directly activate this protein. Knockout studies in mice suggested a critical role of the gene encoding this protein in regulating myocyte apoptosis. Alternatively spliced transcript variants encoding distinct isoforms have been described.
Specifications
| GP130 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| Q00560 | |
| IL6ST | |
| Mouse myeloma cell line NS0-derived recombinant mouse gp130, Gln23-Glu617 (predicted). | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 125604 | |
| Unconjugated | |
| IL6ST | |
| 5133400A03Rik; AA389424; Ac1055; BB405851; CD130; CD130 antigen; CDW130; D13Ertd699e; EGK_16499; Gp130; gp130 of the rheumatoid arthritis antigenic peptide-bearing soluble form; gp130, oncostatin M receptor; GP130-RAPS; IL-6; IL-6 receptor subunit beta; IL-6R subunit beta; IL-6RB; IL-6R-beta; IL6R-beta; IL6ST; interleukin 6 signal transducer; interleukin 6 signal transducer (gp130, oncostatin M receptor); interleukin 6 signal transducer receptor; interleukin receptor beta chain; interleukin-6 receptor subunit beta; Interleukin-6 signal transducer; interleukin-6 signal transducer receptor; membrane glycoprotein; membrane glycoprotein 130; membrane glycoprotein gp130; oncostatin-M receptor subunit alpha | |
| Rat | |
| Protein A/G | |
| RUO | |
| 16195 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction