missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GP2 Antibody (GP2/1803), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75779-20ug
This item is not returnable.
View return policy
Description
GP2 Monoclonal antibody specifically detects GP2 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
| GP2 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 2-4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| P55259 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 2813 | |
| Store at 4°C. | |
| IgG2a κ |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array | |
| GP2/1803 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| DKFZp779K0533, glycoprotein 2 (zymogen granule membrane), pancreatic secretory granule membrane major glycoprotein GP2, pancreatic zymogen granule membrane associated protein GP2, Pancreatic zymogen granule membrane protein GP-2, ZAP75 | |
| Recombinant fragment of human GP2 protein (around aa 35-179) (exact sequence is proprietary) (Uniprot: P55259) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction