missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GP340 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5149092
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
Glycoprotein-340 is member of the scavenger receptor superfamily and has been identified as a putative SP-D receptor. Gp-340 has been shown to bind SP-D in a calcium dependent manner through a protein-protein interaction. Like SP-D, gp-340 is mainly found in lung tissue and immunohistochemistry revealed high expression in alveolar macrophages. An identical protein is present in saliva (salivary agglutinin) and both forms are encoded by the gene DMBT1.
Specifications
| GP340 | |
| Polyclonal | |
| Unconjugated | |
| DMBT1 | |
| apactin; CRP; CRP-[a]; CRP-[b]; Crpd; CRP-ductin; DBMT1; deleted in malignant brain tumors 1; deleted in malignant brain tumors 1 protein; deleted in malignant brain tumors 1 pseudogene; DMBT1; Ebnerin; glycoprotein 300; Glycoprotein 340; gp300; GP340; gp-340; hensin; Mucin-like glycoprotein; muclin; p80; pancrin; RP11-481L19.1; SAG; Salivary agglutinin; surfactant pulmonary-associated D-binding protein; Vomeroglandin | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 12945 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| Q60997 | |
| DMBT1 | |
| E. coli-derived recombinant mouse DMBT1, Trp215-Gly420. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction