missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ GPR3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30864-100ug
This item is not returnable.
View return policy
Description
GPR3 Polyclonal antibody specifically detects GPR3 in Human samples. It is validated for ELISA,Western Blot
Specifications
| GPR3 | |
| Polyclonal | |
| Western Blot : 1-5 μg/mL, ELISA | |
| ACCA, G protein-coupled receptor 3 | |
| Synthetic peptide derived from the GPR3 | |
| 100 μg | |
| GPCR | |
| 2827 | |
| Store at -20°C. Avoid freeze/thaw cycles. | |
| IgG |
| ELISA, Western Blot | |
| Unconjugated | |
| Phosphate buffered saline | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction