missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR40 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575351
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
This gene encodes a member of the GP40 family of G protein-coupled receptors that are clustered together on chromosome 19. The encoded protein is a receptor for medium and long chain free fatty acids and may be involved in the metabolic regulation of insulin secretion. Polymorphisms in this gene may be associated with type 2 diabetes.
Specifications
| GPR40 | |
| Polyclonal | |
| Unconjugated | |
| FFAR1 | |
| FFA1R; FFAR; Ffar1; Free fatty acid receptor 1; G protein-coupled receptor 40; G protein-coupled receptor GPR40; GPCR40; GPR40; G-protein coupled receptor 40; HGNC:4498 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 233081, 266607, 2864 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14842, Q76JU9, Q8K3T4 | |
| FFAR1 | |
| Synthetic peptide corresponding to amino acids 201-250 of Human GPR40. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction