missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPR84 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA130069
This item is not returnable.
View return policy
Description
PA1-30069 detects GPCR EX33 from human samples.
G protein-coupled receptors (GPCRs, or GPRs) contain 7 transmembrane domains and transduce extracellular signals through heterotrimeric G proteins. GPCR EX33 is a member of this family (subfamily Orphan A).
Specifications
| GPR84 | |
| Polyclonal | |
| Unconjugated | |
| GPR84 | |
| EX33; G protein-coupled receptor 84; GPCR4; GPR84; G-protein coupled receptor 84; inflammation-related G protein-coupled receptor EX33; inflammation-related G-protein coupled receptor EX33; orphan G protein-coupled receptor 84 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100062469, 53831 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9NQS5 | |
| GPR84 | |
| Synthetic 19 amino acid peptide from 2nd extracellular domain of human GPR84. | |
| 25 μg | |
| Primary | |
| Horse, Human, Monkey | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction