missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR97 Antibody (2435C) [Alexa Fluor™ 532], Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB10097X
This item is not returnable.
View return policy
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF
Specifications
| GPR97 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, CyTOF | |
| 2435C | |
| Western Blot, Flow Cytometry, CyTOF-ready | |
| G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
| Human embryonic kidney cell, HEK293-transfected human GPR97, Met1-Leu249, Accession # Q86Y34 | |
| 0.1 mL | |
| GPCR | |
| 222487 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction