missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GPR97 Antibody (CL11463) [Alexa Fluor™ 647], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-21194AF647
This item is not returnable.
View return policy
Description
GPR97 Monoclonal antibody specifically detects GPR97 in Human samples. It is validated for Western Blot
Specifications
| GPR97 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| G protein-coupled receptor 97, Pb99, PGR26G-protein coupled receptor PGR26, probable G-protein coupled receptor 97 | |
| This antibody was developed using a synthetic peptide derived from Q86Y34, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| GPCR | |
| 222487 | |
| Store at 4⪚C in the dark. | |
| IgG2a |
| Western Blot | |
| CL11463 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction