missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GPX2 Monoclonal Antibody (496010)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA561560
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
This gene is a member of the glutathione peroxidase family and encodes a selenium-dependent glutathione peroxidase that is one of two isoenzymes responsible for the majority of the glutathione-dependent hydrogen peroxide-reducing activity in the epithelium of the gastrointestinal tract. Studies in knockout mice indicate that mRNA expression levels respond to luminal microflora, suggesting a role of the ileal glutathione peroxidases in preventing inflammation in the GI tract.
Specifications
| GPX2 | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P18283, P83645, Q9JHC0 | |
| GPX2 | |
| E. coli-derived recombinant human Glutathione Peroxidase 2, Gly41-Ile190. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG2b |
| Western Blot | |
| 496010 | |
| Unconjugated | |
| GPX2 | |
| gastrointestinal glutathione peroxidase; GI-GPx; Glutathione peroxidase 2; glutathione peroxidase 2 (gastrointestinal); glutathione peroxidase-gastrointestinal; Glutathione peroxidase-related protein 2; GPRP; GPRP-2; GPX2; GPx-2; GPX-GI; GSHPx-2; GSHPX-GI; HGNC:4554; intestinal GPx | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 14776, 2877, 29326 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction