missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Gpx3 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA518677
This item is not returnable.
View return policy
Description
This antibody is predicted to react with canine, human and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Glutathione peroxidases (Gpxs) are ubiquitously expressed proteins which catalyze the reduction of hydrogen peroxides and organic hydroperoxides by glutathione. There are several isoforms which differ in their primary structure and localization. The classical cytosolic/mitochondrial GPx1 (cGPx) is a selenium-dependent enzyme, first of the GPx family to be discovered. GPx2, also known as gastrointestinal GPx (GI-GPx), is an intracellular enzyme expressed only at the epithelium of the gastrointestinal tract. Extracellular plasma GPx (pGPx or GPx3) is mainly expressed by the kidney from where it is released into the blood circulation. Phospholipid hydroperoxide GPx4 (PH-GPx) expressed in most tissues, can reduce many hydroperoxides including hydroperoxides integrated in membranes, hydroperoxy lipids in low density lipoprotein or thymine. All mammalian GPx family members, except for the recently described Cys containing GPx3 and epididymis-specific secretory GPx (eGPx or GPx5) isoforms, possess selenocysteine at the active site (4-5).
Specifications
| Gpx3 | |
| Polyclonal | |
| Unconjugated | |
| Gpx3 | |
| AA960521; EGPx; extracellular glutathione peroxidase; extracellular GPx; glutathione peroxidase 3; glutathione peroxidase 3 (plasma); GPx; Gpx3; GPx-3; Gpxp; GPx-P; GSHPx-3; GSHPx-P; plasma form; plasma glutathione peroxidase; plasma GPx | |
| Goat | |
| Ammonium sulfate precipitation, Antigen affinity chromatography | |
| RUO | |
| 14778, 2878 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunohistochemistry (PFA fixed), Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P22352, P46412 | |
| Gpx3 | |
| Synthetic peptide sequence (NQFGKQEPGENSE) corresponding to the internal amino acids of GPX3 (aa 102-114). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction