missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gr-1/Ly-6G Antibody (1A8), PerCP, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-53131PCP
This item is not returnable.
View return policy
Description
Gr-1/Ly-6G Monoclonal specifically detects Gr-1/Ly-6G in Mouse samples. It is validated for Flow Cytometry.
Specifications
| Gr-1/Ly-6G | |
| Monoclonal | |
| PerCP | |
| PBS with 0.05% Sodium Azide | |
| LY6G | |
| Ly-6G transfected EL-4J cell line. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| 1A8 | |
| Flow Cytometry | |
| C6orf23, G6D, LY6-D, LY6G6D, lymphocyte antigen 6 family member G6D, MEGT1, NG25 | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| 58530 | |
| Store at 4C in the dark. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction