Learn More
GR repeat Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 23978-1-AP-150UL
Description
This antibody is used to detect the GR repeated sequence. The recombit proteins GST-SNRPD1 and 6xHis tag- SNRPD1 which contains the sequence 'GRGRGRGRGRGRGRGRGR' were used to detect this antibody. Both proteins can be recognized by the antibod
The C9orf72 "GGGGCC" repeat sequence codes five repeat peptide "GA repeat; GAGAGAGAGA", "GP repeat; GPGPGPGPG", "GR repeat; GRGRGRGRG", "AP repeat; APAPAPAPA" and "PR repeat; PRPRPRPRP". It was described previously that aggregated forms of poly-GA and poly-GP proteins do not enter the separation gel.Specifications
| GR repeat | |
| Polyclonal | |
| Unconjugated | |
| C9RAN, GGGCC, GR repeat, GRGRGRGRG, GRGRGRGRGR, Poly-GR | |
| Peptide | |
| 150 μL | |
| Primary | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.24 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.