missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ Grade 100/N Smooth Surface Folded Filter Qualitative Filter Papers

Used for routine analyses like clarification and determination of substances

Brand:  Sartorius™ FT-4-328-240

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 10741893

  • 55.90€ / Pack of 100
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

  • Made of refined pulp and cotton linters with an >95% α-cellulose content
  • Ash content between 0.1–0.15% (grade 100/N <0.1%)
  • Wet-strengthened
  • Supplied in folded filters as well as customer-specific cuts
Specifications

Specifications

Folded
30μm/sec.
100/N
0.18 mm
240 mm
Cotton Linters, Refined Pulp
For qualitative-technical analyses, used for routine analyses like clarification, determination of substances, but also as discs with a center hole for technical applications
100/Pk.
Qualitative Filter Paper
240 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions