Learn More
Globe Scientific Graduated Transfer Pipet
Product Code. 31922526
Change view
Click to view available options
Quantity:
100 Pk., 5 Pk./Unit
Unit Size:
Pack of 500
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
31922526 100 Pk., 5 Pk./Unit Pack of 500
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 31922526 Supplier Globe Scientific Supplier No. 137018H
Estimated Shipment: 04-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Graduated transfer pipets are designed for measurements and feature molded-in graduation marks that will never fade or wash off.
  • Molded-in graduation marks to 1 mL that will never fade or wash off
Specifications
Aspirator Plug No
Autoclavable Not Autoclavable
Calibrated No
Material LDPE
Sterility Sterile
Graduated Yes
Graduation Type Molded
Length (Metric) Tip 150 mm
Quantity 100 Pk., 5 Pk./Unit
Packaging Type Individually Wrapped
Color Clear
Volume (Metric) 5 mL
Type Transfer Pipet
Disposable Single-use