Learn More
3M™ Graffiti Remover 3000
Product Code. 17189340
Change view
Click to view available options
Volume (Metric):
500 mL
5 L
Unit Size:
4 bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
17189340 5 L 4 bottles
17179340 500 mL 4 bottles
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 17189340 Supplier 3M™ Supplier No. 7100030786
  • ONLINE EXCLUSIVE PRICE
    924.00€
    570.30€ / Pack of 4
    Save 353.70€
    38% Off
 Restricted Item
Estimated Shipment: 18-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Formulated to quickly and effectively remove graffiti from brickwork and concrete surfaces. Effective on a wide range of graffiti substances including aerosol spray paint, permanent markers, crayon, and emulsion paint.
  • Graffiti remover for plastic, painted, or varnished surfaces
  • Effective against aerosol spray paint, permanent markers, lipstick, crayon, and emulsion paint
  • The solvent’s liquid form allows for quick penetration and rapid breakdown of graffiti
  • Does not contain NEP (N-Ethyl-pyrrolidone) or NMP (N-Methyl-2-pyrrolidone) used in other graffiti removers, which can pose a significant risk to health
  • Has minimal odor to enhance user comfort and efficiency unlike traditional chemical removers which emit powerful odors and harmful vapors
  • Low environmental impact since the product does not contain aromatic, aliphatic, or chlorinated solvents
TRUSTED_SUSTAINABILITY
Specifications
Product Type Graffiti Remover
Volume (Metric) 5 L