missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRAMD1A Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-32148PCP
This item is not returnable.
View return policy
Description
GRAMD1A Polyclonal antibody specifically detects GRAMD1A in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| GRAMD1A | |
| Polyclonal | |
| Western Blot, Immunoprecipitation | |
| FLJ22411, FLJ90346, GRAM domain containing 1A, GRAM domain-containing protein 1A, KIAA1533 | |
| The immunogen this antibody was made to, maps to a region between residue 674 to 724 of human GRAM Domain Containing 1A using the numbering given in entry NP_065946.2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| PerCP | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 57655 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction