missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Granzyme G Monoclonal Antibody (207639)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA560973
This item is not returnable.
View return policy
Description
Reconstitute at 0.5 mg/mL in sterile PBS.
Specifications
| Granzyme G | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with trehalose and no preservative | |
| P13366 | |
| Gzmg | |
| Mouse myeloma cell line NS0-derived recombinant mouse Granzyme G, Ile21-Leu248. | |
| 500 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG2a |
| Western Blot | |
| 207639 | |
| Unconjugated | |
| Gzmg | |
| AKA granzyme G; CTL serine protease 1; CTL serine protease 3; Ctla7; Ctla-7; granzyme G; GrzG; Gzmg; MCSP-1 | |
| Rat | |
| Protein A/G | |
| RUO | |
| 14944 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction