Learn More
Greiner Bio-One Serum Tubes: Clot Activator with Gel Separator
Product Code. 15422530
Change view
Click to view available options
Capacity (Metric):
2.5 mL
3.5 mL
5 mL
8 mL
13 x 100 mm
Closure Color:
Gold
Red with White Ring
Red with Yellow Ring
Unit Size:
Pack of 100
Pack of 1200
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Closure Color unitSize
15422530 3.5 mL Red with Yellow Ring Pack of 50
15452550 5 mL Gold Pack of 100
13008652 3.5 mL Red with Yellow Ring Pack of 1200
15422550 8 mL Red with Yellow Ring Pack of 50
10144534 8 mL Red with Yellow Ring Pack of 1200
15484641 13 x 100 mm Red with Yellow Ring Pack of 1200
15402560 13 x 100 mm Red with Yellow Ring Pack of 50
13068682 2.5 mL Red with White Ring Pack of 1200
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 15422530 Supplier Greiner Bio-One Supplier No. 454067
 Restricted Item
In Stock
Add to basket
Edge
Add to basket
For routine clinical chemistry, immunohematology, serology or immunology testing
  • Coated with micronized silica particles which activate clotting when tubes are gently inverted
  • Tubes with gel contain a barrier gel on the tube bottom that during centrifugation moves upward to the serum-clot interface to form a stable barrier, separating serum from cells
  • Clear as glass with equal or better performance
  • Molded to be virtually unbreakable, providing a better level of safety than glass
  • Tubes have an 18-month shelf life from the date of manufacture
  • Pediatric tubes come with white rings for quick identification
  • Safety Twist Caps can be removed and replaced several times
  • Nonscrew caps can be removed and replaced as often as required
Compliance
Tubes with gel separator are FDA 510(k) cleared for therapeutic drug monitoring (TDM)
TRUSTED_SUSTAINABILITY
Specifications
Additive Microscopic Silica Particles with Separation Gel
Material Plastic
Closure Type Non-ridged Pull Cap
Capacity (Metric) 3.5 mL
Closure Color Red with Yellow Ring