missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GREM2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13892-1-AP-20UL
This item is not returnable.
View return policy
Description
GREM2 encodes a member of the BMP (bone morphogenic protein) antagonist family. Like BMPs, BMP antagonists contain cystine knots and typically form homo- and heterodimers. The CAN (cerberus and dan) subfamily of BMP antagonists, to which this gene belongs, is characterized by a C-terminal cystine knot with an eight-membered ring. The antagonistic effect of the secreted glycosylated protein encoded by this gene is likely due to its direct binding to BMP proteins. As an antagonist of BMP, this gene may play a role in regulating organogenesis, body patterning, and tissue differentiation.Specifications
| GREM2 | |
| Polyclonal | |
| Unconjugated | |
| GREM2 | |
| CKTSF1B2, DAN domain family member 3, DAND3, GREM2, Gremlin 2, PRDC | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23893, 64388 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.59 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O88273, Q9H772 | |
| GREM2 | |
| GREM2 Fusion Protein Ag4854 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction