missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RCAN1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98713AF594
This item is not returnable.
View return policy
Description
RCAN1 Polyclonal antibody specifically detects RCAN1 in Human samples. It is validated for Western Blot
Specifications
| RCAN1 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Alexa Fluor 594 | |
| ADAPT78, calcipressin-1, calcium and oxidant-inducible mRNA, Down syndrome candidate region 1, Down syndrome critical region gene 1, Down syndrome critical region protein 1, DSC1, modulatory calcineurin-interacting protein 1, Myocyte-enriched calcineurin-interacting protein 1, near DSCR proline-rich protein, regulator of calcineurin 1CSP1DSCR1MCIP1RCN1 | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the center region of the Human RCAN1. | |
| 0.1 mL | |
| Cancer, Neuroscience, Signal Transduction | |
| 1827 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction