missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRHL2 Antibody (CL3760) [CoraFluor™ 1], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61634CL1
This item is not returnable.
View return policy
Description
GRHL2 Monoclonal antibody specifically detects GRHL2 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| GRHL2 | |
| Monoclonal | |
| CoraFluor 1 | |
| BOMMGC149294, Brother of mammalian grainyhead, deafness, autosomal dominant 28, DFNA28, FLJ13782, grainyhead-like 2 (Drosophila), grainyhead-like protein 2 homolog, TFCP2L3MGC149295, Transcription factor CP2-like 3FLJ11172 | |
| This antibody was developed using a recombinant protein derived from Q6ISB3, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL3760 | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 79977 | |
| Store at 4⪚C in the dark. Do not freeze. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction