Learn More
Kinematica™ Stainless Steel Mixing attachment for MB 800 Laboratory Mixer
Product Code. 11999058 Shop All Kinematica Products
Change view
Click to view available options
Capacity (Metric):
2000 mL
4000 mL
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
11999058 4000 mL Each
11989058 2000 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11999058 Supplier Kinematica™ Supplier No. 36050055
  • ONLINE EXCLUSIVE PRICE
    1275.75€
    1184.00€ / Pack of 1
    Save 91.75€
    7% Off
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Mixing attachment for MB 800 consists of stainless steel, gasket and blade head as well as lid with opening for material addition.
Specifications
Capacity (Metric) 4000 mL
Blade Material Stainless Steel
For Use With (Equipment) MB 800