missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRIP1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
195.00€
Specifications
| Antigen | GRIP1 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL |
| Applications | Immunocytochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18527451
|
Novus Biologicals
NB120-2842-0.025mg |
0.025 mg | |||||||
Beskrivelse
GRIP1 Polyclonal specifically detects GRIP1 in Human, Hamster samples. It is validated for Immunocytochemistry/Immunofluorescence.Tekniske data
| GRIP1 | |
| Immunocytochemistry/Immunofluorescence 1 - 2 μg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| glutamate receptor interacting protein 1, glutamate receptor-interacting protein 1, GRIP, GRIP-1 | |
| GRIP1 | |
| IgG | |
| Immunogen affinity purified | |
| GRIP1 |
| 1 mg/mL | |
| Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Cancer, Neuronal Cell Markers, Neurotransmission, Transcription Factors and Regulators | |
| PBS with 0.05% Sodium Azide | |
| 23426 | |
| Synthetic peptide corresponding to residues P(206) L P D S E E E G H D S(217) C of human GRIP1. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
For Research Use Only
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel