missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ GRK5 Polyclonal Antibody

Goat Polyclonal Antibody
Brand: Invitrogen™ PA5148864
This item is not returnable.
View return policy
Description
Reconstitute at 0.2 mg/mL in sterile PBS.
GRK5 is a serine/threonine kinase that regulates the activity of a variety of G protein-coupled receptors and the motility of polymorphonuclear leukocytes. The protein phosphorylates the activated forms of G protein-coupled receptors thus initiating their deactivation. It has also been shown to play a role in regulating the motility of polymorphonuclear leukocytes.
Specifications
| GRK5 | |
| Polyclonal | |
| Unconjugated | |
| GRK5 | |
| 1200003L19Rik; Alpha-CP4; AlphaCP-4; FP2025; G protein-coupled receptor kinase 5; G protein-coupled receptor kinase GRK5; Gprk5; GRK; Grk5; GRK5 protein kinase; GRK-pan; Pan-GRK; Pcbp4; poly(rC) binding protein 4; poly(rC)-binding protein 4 | |
| Goat | |
| Antigen affinity chromatography | |
| RUO | |
| 11476, 14773, 236300, 2869, 59075, 59092 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Lyophilized |
| Western Blot | |
| 0.2 mg/mL | |
| PBS with trehalose and no preservative | |
| P34947, Q62833, Q8VEB1 | |
| GRK5 | |
| E. coli-derived recombinant human GRK5, Pro463-Ser590. | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction