missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GRPEL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12720-1-AP-150UL
This item is not returnable.
View return policy
Description
GRPEL1 (GrpE Like 1, Mitochondrial) is a Protein Coding gene. Diseases associated with GRPEL1 include Human Monocytic Ehrlichiosis and Lymphangitis. Among its related pathways are Metabolism of proteins and Mitochondrial protein import. Gene Ontology (GO) annotations related to this gene include protein homodimerization activity and chaperone binding. An important paralog of this gene is GRPEL2.Specifications
| GRPEL1 | |
| Polyclonal | |
| Unconjugated | |
| GRPEL1 | |
| GREPEL1, GRPEL1, HMGE, Mt GrpE1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 17713, 79563, 80273 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P97576, Q99LP6, Q9HAV7 | |
| Grpel1 | |
| GRPEL1 Fusion Protein Ag3415 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction