missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ GSK 2193874
GREENER_CHOICE

Product Code. 18757646
Change view
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18757646 25 mg 25mg
18750809 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18757646 Supplier Tocris Bioscience™ Supplier No. 5106/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Potent and selective TRPV4 antagonist; orally active

GSK 2193874 is a potent and selective TRPV4 antagonist (IC50 values are 2 and 40 nM for rat and human receptors, respectively); inhibits Ca2+ influx through TRPV4 channels. Prevents and reverses pulmonary edema after myocardial infarction in vivo models. Selective over a panel of ∽200 human receptors, channels and enzymes. Orally active.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-([1,4'-Bipiperidin]-1'-ylmethyl)-7-bromo-N-(1-phenylcyclopropyl)-2-[3-(trifluoromethyl)phenyl]-4-quinolinecarboxamide
CAS 1336960-13-4
Assay Percent Range >98%
Quantity 25 mg
Molecular Formula C37H38BrF3N4O
Formula Weight Observed MW: 691.62
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.