Specifications
| Chemical Name or Material | GSK-5959 |
| CAS | 901245-65-6 |
| Synonym | N-[2,3-Dihydro-1,3-dimethyl-2-oxo-6-(1-piperidinyl)-1 H-benzimidazol-5-yl]-2-methoxy-benzamide |
| Quantity | 100 mg |
| Molecular Formula | C22 H26 N4 O3 |
| Formula Weight | 394.47 |
| Physical Form | Solid |
| Product Type | Reagent |
| Purity | 95% |