missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK3A Rabbit anti-Human, Mouse, Rat, Zebrafish, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | GSK3A |
|---|---|
| Concentration | 0.39 mg/mL |
| Applications | Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16881624
|
Proteintech
13419-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16871624
|
Proteintech
13419-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Glycogen Synthase Kinase-3 differs from most serine/threonine kinases in that it is active in the absence of the action of signaling pathways. Two isoforms exist, GSK-3alpha and GSK-3beta. The function of GSK-3 is to phosphorylate glycogen synthase and thereby inactivate it. Insulin action stimulates the PI 3-kinase pathway, resulting in Akt activation, which phosphorylates and inactivates GSK-3.Specifications
| GSK3A | |
| Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Zebrafish, Rat | |
| P18265, P49840, Q2NL51 | |
| 2931, 30664, 50686, 606496 | |
| GSK3A Fusion Protein Ag4168 | |
| Primary | |
| -20°C | |
| GSK3A |
| 0.39 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GSK3 alpha, GSK3A | |
| Gsk3a, gsk3ab | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title