missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSK3B Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
145.00€ - 410.00€
Specifications
| Antigen | GSK3B |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16862715
|
Proteintech
24198-1-AP-20UL |
20 μL |
145.00€
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16852715
|
Proteintech
24198-1-AP-150UL |
150 μL |
410.00€
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Menin is a putative tumor suppressor associated with a syndrome known as multiple endocrine neoplasia type 1. In vitro studies have shown menin is localized to the nucleus, possesses two functional nuclear localization signals, and inhibits transcriptional activation by JunD, however, the function of this protein is not known. Two messages have been detected on northern blots but the larger message has not been characterized. Two variants of the shorter transcript have been identified where alternative splicing affects the CDS. Five variants where alternative splicing takes place in the 5′ UTR have also been identified.Specifications
| GSK3B | |
| Immunofluorescence, Flow Cytometry, Immunohistochemistry (Paraffin), Immunocytochemistry, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Mouse, Human | |
| P49841, Q9WV60 | |
| 2932, 56637 | |
| GSK3B Fusion Protein Ag21438 | |
| Primary | |
| -20°C | |
| GSK3B |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| GSK 3 beta, GSK3 beta, GSK3A B, GSK3B, GSK-3B, GSK3B 216P, gsk3beta | |
| GSK3B | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title