missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSPT1 Antibody (CL13332), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-18565-25ul
This item is not returnable.
View return policy
Description
GSPT1 Monoclonal antibody specifically detects GSPT1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| GSPT1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| CL13332 | |
| Western Blot 1 μg/ml, Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1 | |
| This antibody was developed against a sythetic peptide for P15170. Actual immunogen sequence is proprietary. | |
| 25 μg | |
| Cell Cycle and Replication | |
| 2935 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction