Learn More
GSPT1 Antibody (CL13332) [PE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-18565PE
Product Code. 30293416
  • 770.00€ / 0.10mL
Description
GSPT1 Monoclonal antibody specifically detects GSPT1 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GSPT1
Monoclonal
PE
ERF3A, eRF3aFLJ38048, ETF3A, eukaryotic peptide chain release factor GTP-binding subunit ERF3A, Eukaryotic peptide chain release factor subunit 3a, FLJ39067, G1 to S phase transition 1,551G9.2, G1 to S phase transition protein 1 homolog, GST1
This antibody was developed using a synthetic peptide derived from P15170, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Cycle and Replication
2935
Store at 4⪚C in the dark.
IgG2a
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL13332
PBS
Mouse
Protein A purified