Learn More
GSTM3 Mouse anti-Human, Unconjugated, Clone: 1E2B8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67634-1-IG-150UL
 View more versions of this product
Product Code. 16810376
  • 410.00€ / 150µL
Description
The GSTM3 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GSTM3. This antibody recognizes human, pig antigen. The GSTM3 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
GSTM3
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Pig
Western Blot, ELISA
1E2B8
Unconjugated
150 μL
2947