missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTT1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15838-1-AP-20UL
This item is not returnable.
View return policy
Description
Glutathione S-transferase (GST) theta 1 (GSTT1) is a member of a superfamily of proteins that catalyze the conjugation of reduced glutathione to a variety of electrophilic and hydrophobic compounds. Human GSTs can be divided into five main classes: alpha, mu, pi, theta, and zeta. The theta class includes GSTT1 and GSTT2. The GSTT1 and GSTT2 share 55% amino acid sequence identity and both of them were claimed to have an important role in human carcinogenesis. The GSTT1 gene is located approximately 50kb away from the GSTT2 gene. The GSTT1 and GSTT2 genes have a similar structure, being composed of five exons with identical exon/intron boundaries.Specifications
| GSTT1 | |
| Polyclonal | |
| Unconjugated | |
| GSTT1 | |
| Glutathione transferase T1 1, GST class theta 1, GSTT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 14871, 25260, 2952 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P30711, Q01579, Q64471 | |
| Gstt1 | |
| GSTT1 Fusion Protein Ag8588 | |
| 20 μL | |
| Primary | |
| Rat, Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction