missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GSTZ1 Antibody (24E9.G9.G8), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-44177AF700
This item is not returnable.
View return policy
Description
GSTZ1 Monoclonal antibody specifically detects GSTZ1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| GSTZ1 | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| 2954 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| 24E9.G9.G8 | |
| Western Blot, ELISA, Immunohistochemistry | |
| EC 2.5.1.18, EC 5.2.1.2, glutathione S-alkyltransferase, glutathione S-aralkyltransferase, Glutathione S-transferase zeta 1, glutathione transferase zeta 1, GSTZ1-1maleylacetoacetate isomerase, MAAIglutathione S-aryltransferase, MAI, maleylacetone isomerase, MGC2029, S-(hydroxyalkyl)glutathione lyase | |
| This Protein A purified GSTZ1 Antibody (24E9.G9.G8) was prepared by repeated immunizations in mice with GSTGSTZ1 Protein (Uniprot: O43708) | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction