missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GTF2H2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16005-1-AP-20UL
This item is not returnable.
View return policy
Description
This is a rabbit polyclonal antibody produced by part of the N-terminal region of human GTF2H2.
Initiation of transcription from protein-coding genes in eukaryotes is a complex process that requires RNA polymerase II, as well as families of basal transcription factors. Binding of the factor TFIID (TBP) to the TATA box is believed to be the first step in the formation of a multiprotein complex containing several additional factors, including TFIIA, TFIIB, TFIIE, TFIIF and TFII. TFIIH (or BTF2) is a multisubunit transcription/DNA repair factor that possesses several enzymatic activities. The core of TFIIH is composed of five subunits, designated p89 (XPB or ERCC3), p62, p52, p44 and p34. Additional subunits of the TFIIH complex are p80 (XPD or ERCC2) and the ternary kinase complex composed of Cdk7, cyclin H and MAT1. Both p89 and p80 have ATP-dependent helicase activity. The p62, p52 and p44 subunits have been shown to be involved in nucleotide excision repair.Specifications
| GTF2H2 | |
| Polyclonal | |
| Unconjugated | |
| GTF2H2 | |
| BTF2, BTF2 p44, BTF2P44, GTF2H2, T BTF2P44, TFIIH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23894, 294693, 2966 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.29 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A0JN27, Q13888, Q9JIB4 | |
| Gtf2h2 | |
| GTF2H2 Fusion Protein Ag8653 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction