Learn More
GTF2H3 Monoclonal Antibody (OTI4B5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812723S
Description
GTF2H3 Monoclonal antibody specifically detects GTF2H3 in Human samples. It is validated for Western BlotSpecifications
| GTF2H3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O43364 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 3199 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI4B5 | |
| Unconjugated | |
| HOXA2 | |
| 34kDa; 5033417D07Rik; Basic transcription factor 2 34 kDa subunit; BTF2; BTF2 p34; BTF2-p34; C730029A10; D5Ertd679e; General transcription factor IIH polypeptide 3; general transcription factor IIH subunit 3; general transcription factor IIH, polypeptide 3; general transcription factor IIH, polypeptide 3, 34kDa; Gtf2h3; P34; TFB4; TFIIH; TFIIH basal transcription factor complex p34 subunit | |
| Human recombinant protein fragment corresponding to amino acids 57-319 of human HOXA2 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.