missing translation for 'onlineSavingsMsg'
Learn More

Guanylyl Cyclase alpha 1 Antibody (SR2180), Novus Biologicals™

Rabbit Monoclonal Antibody

338.00€ - 449.00€

Specifications

Antigen Guanylyl Cyclase alpha 1
Clone SR2180
Dilution Western Blot 1:500-1:2000
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18650755
View Documents
Novus Biologicals
NBP3-22388-100ul
100 μg
449.00€
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18633194
View Documents
Novus Biologicals
NBP3-22388-50ul
50 μg
338.00€
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Guanylyl Cyclase alpha 1 Monoclonal antibody specifically detects Guanylyl Cyclase alpha 1 in Human samples. It is validated for Western Blot
Specifications

Specifications

Guanylyl Cyclase alpha 1
Western Blot 1:500-1:2000
Monoclonal
Purified
RUO
Human
EC 4.6.1.2, GC-S-alpha-1, GCS-alpha-1, GCS-alpha-3, guanylate cyclase 1, soluble, alpha 3, guanylate cyclase soluble subunit alpha-3, GUC1A3GUCA3, GUCSA3GC-SA3, GUCY1A1, Soluble guanylate cyclase large subunit
A synthesized peptide derived from human Guanylyl Cyclase alpha 1 (Uniprot #: Q02108)
Primary
Store at -20° C. Avoid freeze/thaw cycles.
SR2180
Western Blot
Unconjugated
Rabbit
Signal Transduction, Vision
PBS, pH 7.4, 150mM NaCl, 50% glycerol.
2982
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.